MilliporeSigma
  • Efficacy of a 5-day extended therapy program during lactation with cephapirin sodium in dairy cows chronically infected with Staphylococcus aureus.
Efficacy of a 5-day extended therapy program during lactation with cephapirin sodium in dairy cows chronically infected with Staphylococcus aureus.
The Canadian veterinary journal = La revue veterinaire canadienne (2010-03-02)
Jean-Philippe Roy, Luc DesCôteaux, Denis DuTremblay, Francis Beaudry, Johanne Elsener
ABSTRACT
This study determined the efficacy of a 5-day extended therapy with cephapirin sodium in dairy cows chronically infected with Staphylococcus aureus. Chronically infected cows selected from 14 dairy herds in the St-Hyacinthe region, Québec were randomly allocated to a group of 31 cows treated for 5 consecutive days with 200 mg of cephapirin per quarter BID or a group of 30 untreated control cows. Bacteriological cure was determined by 3 negative bacterial cultures at 10, 24, and 31 days after treatment. The cow cure rates were 25.8% (8/31) in the treated cows and 3.3% (1/30) in the control group (P = 0.013). The quarter cure rates at first sampling post-treatment were 77.6% (38/49) and 18% (9/50) in the treated and the control groups, respectively (P < 0.0001). A 5-day extended therapy with cephapirin is effective in treating cows chronically infected with S. aureus.
Product Description
Supelco
Cefapirin sodium, analytical standard