- Influence of polymers ratio on insulin-loaded nanoparticles based on poly-epsilon-caprolactone and Eudragit RS for oral administration.
Influence of polymers ratio on insulin-loaded nanoparticles based on poly-epsilon-caprolactone and Eudragit RS for oral administration.
Drug delivery (2009-10-21)
M Socha, A Sapin, C Damgé, P Maincent
PMID19839787
ABSTRACT
Nanoparticles loaded with two different commercial insulins (Actrapid, Novorapid and based on different blends of a biodegradable polyester (poly-epsilon-caprolactone) and a polycationic non-biodegradable acrylic polymer (Eudragit RS) were characterized in vitro. The zeta potential was positive whenever Eudragit RS was part of the nanoparticles matrix. The encapsulation efficiency was ~ 96% except for Novorapid-loaded particles of poly-epsilon-caprolactone (only 35%). In vitro release studies revealed a burst release from nanoparticles, which may be of interest for oral delivery. Novorapid-loaded nanoparticles were orally administered to diabetic rats and allowed the glycemia to be decreased when compared with free nanoparticles.