Merck
  • Monochromatic excimer light 308 nm in monotherapy and combined with topical khellin 4% in the treatment of vitiligo: a controlled study.
Monochromatic excimer light 308 nm in monotherapy and combined with topical khellin 4% in the treatment of vitiligo: a controlled study.
Dermatologic therapy (2009-07-08)
Rosita Saraceno, Steven P Nisticò, Elisabetta Capriotti, Sergio Chimenti
ABSTRACT
Vitiligo is an acquired depigmentation disorder affecting 1-4% of the world's population. Conventional therapies include steroids, photosensitive topical agents, surgical treatments, and phototherapy. The aim of the study was to evaluate the efficacy of monochromatic excimer light 308 nm (MEL), both as a monotherapy and in combination with khellin 4% ointment in vitiligo. Forty-height patients (36 male and 12 female) affected with vitiligo were enrolled in this open prospective study. Patients were selected and divided into three groups: group I included 16 patients treated with MEL 308 nm once-weekly and oral vitamin E; group II included 16 patients treated with MEL 308 nm once-weekly combined with khellin 4% ointment (MEL-K) and oral vitamin E; group III (control group) included 16 patients treated only with oral vitamin E. Efficacy was assessed at the end of 12 weeks based on the percentage of repigmentation. Group I (MEL-group) showed a moderate repigmentation in 2/16 (12.5%) patients, good repigmentation in 10/16 (62.5%), and excellent repigmentation in 4/16 (25%) patients. Group II (MEL-K group) presented moderate repigmentation in 2/16 (12.5%) patients, good repigmentation in 5/16 (31.25%), and excellent repigmentation in 9/16 (56.25%). Group III (control group) showed a moderate repigmentation in 3/16 patients (18.75%), a good repigmentation in 1/16 (6.25%) patient, while 10/16 (62.5%) patients did not show signs of repigmentation. The clinical response achieved in group I and II was higher compared with group III (control group) without showing significant differences. MEL 308 nm, alone and/or combined with khellin 4% offered encouraging results and it may be considered a valid therapeutic option worthy of consideration in the treatment of vitiligo.
Sigma-Aldrich
Khellin, for microscopy