- Diurnal changes in the synthesis of the neurosteroid 7alpha-hydroxypregnenolone stimulating locomotor activity in newts.
Diurnal changes in the synthesis of the neurosteroid 7alpha-hydroxypregnenolone stimulating locomotor activity in newts.
Annals of the New York Academy of Sciences (2009-05-22)
Teppei Koyama, Shogo Haraguchi, Hubert Vaudry, Kazuyoshi Tsutsui
PMID19456382
ABSTRACT
We recently identified 7alpha-hydroxypregnenolone as a novel amphibian neurosteroid stimulating locomotor activity in newts. Because male newts show marked diurnal changes in locomotor activity, we hypothesized that 7alpha-hydroxypregnenolone may be a key factor for the induction of diurnal changes in locomotor activity in male newts. In this study, we found diurnal changes in 7alpha-hydroxypregnenolone synthesis in the brain of male newts, which paralleled locomotor activity. Interestingly, the production of 7alpha-hydroxypregnenolone in the male newt brain increased during the dark phase when locomotor activity of males was high.