Merck
  • Is toluene diamine a sensitizer and is there cross-reactivity between toluene diamine and toluene diisocyanate?
Is toluene diamine a sensitizer and is there cross-reactivity between toluene diamine and toluene diisocyanate?
Toxicological sciences : an official journal of the Society of Toxicology (2009-04-01)
Jeroen A J Vanoirbeek, Vanessa De Vooght, Nicholas Synhaeve, Benoit Nemery, Peter H M Hoet
ABSTRACT
Toluene diamine (TDA) is formed when toluene diisocyanate (TDI), a potent sensitizer, comes in contact with an aqueous environment. The sensitizing capacity of TDA and the cross-reactivity between TDI and TDA are unknown. TDA (5-25%) and TDI (0.3%), dissolved in acetone/olive oil (AOO) (4:1) were tested in the mouse local lymph node assay (LLNA). To determine the capacity of TDA to elicit an asthmatic response and to determine the cross-reaction with TDI, a locally developed experimental mouse model of chemical-induced asthma was used. On days 1 and 8, BALB/c mice received 20 microl of TDI (0.3%), TDA (20%), or AOO (4:1) on each ear. On day 15, they received an intranasal instillation of TDI (0.1%), TDA (0.5%) or AOO (3:2). The EC(3) of TDA in the LLNA is 19%. In the model of chemical-induced asthma, TDI induced a ventilatory response [increased Penh after challenge; increased airway hyperreactivity (AHR)], inflammatory changes (bronchoalveolar lavage neutrophils), and immunological changes (increased CD19(+) lymphocytes, IL-4 and total serum IgE), whereas TDA did not show any of these responses. Mice sensitized with TDI and challenged with TDA also did not show any airway or inflammatory response, although they had increased levels of total serum IgE. Mice sensitized with TDA and challenged with TDI did not show any response. According to the classification of sensitizers in the LLNA, TDA is a weak dermal sensitizer. In the experimental mouse model of chemical-induced asthma, TDA does not act as a respiratory sensitizer, at the concentration used. No cross-reactivity between TDI and TDA was found.
Sigma-Aldrich
2,4-Diaminotoluene, 98%