Merck
  • Is paromomycin an effective and safe treatment against cutaneous leishmaniasis? A meta-analysis of 14 randomized controlled trials.
Is paromomycin an effective and safe treatment against cutaneous leishmaniasis? A meta-analysis of 14 randomized controlled trials.
PLoS neglected tropical diseases (2009-02-18)
Dae Hyun Kim, Hye Jin Chung, Joachim Bleys, Reza F Ghohestani
ABSTRACT
High cost, poor compliance, and systemic toxicity have limited the use of pentavalent antimony compounds (SbV), the treatment of choice for cutaneous leishmaniasis (CL). Paromomycin (PR) has been developed as an alternative to SbV, but existing data are conflicting. We searched PubMed, Scopus, and Cochrane Central Register of Controlled Trials, without language restriction, through August 2007, to identify randomized controlled trials that compared the efficacy or safety between PR and placebo or SbV. Primary outcome was clinical cure, defined as complete healing, disappearance, or reepithelialization of all lesions. Data were extracted independently by two investigators, and pooled using a random-effects model. Fourteen trials including 1,221 patients were included. In placebo-controlled trials, topical PR appeared to have therapeutic activity against the old world and new world CL, with increased local reactions, when used with methylbenzethonium chloride (MBCL) compared to when used alone (risk ratio [RR] for clinical cure, 2.58 versus 1.01: RR for local reactions, 1.60 versus 1.07). In SbV-controlled trials, the efficacy of topical PR was not significantly different from that of intralesional SbV in the old world CL (RR, 0.70; 95% confidence interval, 0.26-1.89), whereas topical PR was inferior to parenteral SbV in treating the new world CL (0.67; 0.54-0.82). No significant difference in efficacy was found between parenteral PR and parenteral SbV in the new world CL (0.88; 0.56-1.38). Systemic side effects were fewer with topical or parenteral PR than parenteral SbV. Topical PR with MBCL could be a therapeutic alternative to SbV in selected cases of the old world CL. Development of new formulations with better efficacy and tolerability remains to be an area of future research.
Sigma-Aldrich
Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)
Sigma-Aldrich
Benzethonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Hyamine® 1622 solution, 4 mM in H2O