Merck
  • Polyphosphoric acid treatment promotes bone regeneration around titanium implants.
Polyphosphoric acid treatment promotes bone regeneration around titanium implants.
Journal of oral rehabilitation (2009-02-13)
K Maekawa, K Shimono, M Oshima, Y Yoshida, B Van Meerbeek, K Suzuki, T Kuboki
ABSTRACT
This study was conducted to evaluate the effect of polyphosphoric acid (PPA) treatment on bone regeneration around titanium (Ti) implants in vivo. Adsorption of PPA by Ti was achieved by immersing Ti implants (2 mm in diameter, 4 mm in length) in different concentrations of PPA solution (0, 1 and 10 wt%) for 24 h at 37 degrees C after proper Ti surface cleaning. The treated Ti implants were implanted on 8-week-old-male rat (n = 30) tibiae. Two or four weeks after implantation, all animals were deeply anaesthetized and underwent perfusion fixation. Ten specimens in each condition were further immersed in the same fixative for 1 week and eventually embedded in polyester resin. Afterwards, undecalcified sections were ground to a thickness of approximately 70 microm parallel to the long axis of the implant. The sections were stained with basic fuchsine and methylene blue and then examined by light microscopy. For quantitative evaluation of bone regeneration around the implants, the bone-implant contact ratio (BICR) was determined. Polyphosphoric acid treatment of the Ti implant surface significantly enhanced direct bone contact to the Ti surface. Especially, the BICRs of the 1 wt% PPA-treated Ti implants were significantly higher than those of the control untreated Ti implants, both 2 and 4 weeks after implantation. At 4 weeks, 10 wt% PPA-treated implants also significantly increased the BICR as compared to that of the untreated Ti implants. These results suggest that PPA treatment promotes osteoconductivity of Ti in vivo.
Sigma-Aldrich
Polyphosphoric acid, reagent grade, 115% H3PO4 basis
Sigma-Aldrich
Polyphosphoric acid, ~105% H3PO4 basis