Merck
  • Efficient dehydrative sialylation of C-4-aminated sialyl-hemiketal donors with Ph2SO/Tf2O.
Efficient dehydrative sialylation of C-4-aminated sialyl-hemiketal donors with Ph2SO/Tf2O.
The Journal of organic chemistry (2009-01-20)
Deju Ye, Wenfeng Liu, Dengyou Zhang, Enguang Feng, Hualiang Jiang, Hong Liu
ABSTRACT
An efficient approach to the dehydrative sialylation of various substrates with C-4-aminated sialyl-hemiketal donors by using the reagent combination of diphenyl sulfoxide and triflic anhydride is reported. By using a C-4-hindered non-nucleophilic amine auxiliary, excellent yields and high alpha-stereoselectivities were obtained for coupling with a wide range of primary and secondary acceptors.
Sigma-Aldrich
Diphenyl sulfoxide, 96%