Merck
  • Rational design of novel, potent piperazinone and imidazolidinone BACE1 inhibitors.
Rational design of novel, potent piperazinone and imidazolidinone BACE1 inhibitors.
Bioorganic & medicinal chemistry letters (2008-05-13)
J N Cumming, T X Le, S Babu, C Carroll, X Chen, L Favreau, P Gaspari, T Guo, D W Hobbs, Y Huang, U Iserloh, M E Kennedy, R Kuvelkar, G Li, J Lowrie, N A McHugh, L Ozgur, J Pan, E M Parker, K Saionz, A W Stamford, C Strickland, D Tadesse, J Voigt, L Wang, Y Wu, L Zhang, Q Zhang
ABSTRACT
Guided by structure-based design, we synthesized two novel series of potent inhibitors of BACE1 and generated extensive SAR around both the prime and non-prime side binding pockets. The key feature of both series is a cyclic amine motif specifically crafted to achieve interactions with both the flap and with the S2' pocket.
Sigma-Aldrich
2-Imidazolidone, 96%