Merck
  • Reproductive and developmental toxicity screening test of tetrahydrofurfuryl alcohol in rats.
Reproductive and developmental toxicity screening test of tetrahydrofurfuryl alcohol in rats.
Reproductive toxicology (Elmsford, N.Y.) (2008-01-15)
Mutsuko Hirata-Koizumi, Atsushi Noda, Akihiko Hirose, Eiichi Kamata, Makoto Ema
ABSTRACT
Twelve male and female rats per group were given tetrahydrofurfuryl alcohol (THFA) by gavage at 0, 15, 50, 150 or 500 mg/kg/day. Males were dosed for 47 days, beginning 14 days before mating, and females were dosed for 42-52 days beginning 14 days before mating to day 4 of lactation throughout the mating and gestation period. Changes in locomotor activity, inhibition of body weight gain, and/or histopathological changes in the thymus, spleen, testes and/or epididymides were observed in males and females at 150 mg/kg and above. No effects of THFA were found on the copulation index, fertility index, or the number of corpora lutea and implantations in pregnant females. At 500 mg/kg, no pregnant females delivered any pups. At 150 mg/kg, gestation length was prolonged, and the total number of pups born and the number of live pups on postnatal days 0 and 4 was markedly decreased. No effects of THFA were found on the sex ratio and body weight of live pups, or the incidence of pups with malformations or variations. Based on these findings, the NOAELs for parental and reproductive/developmental toxicity of THFA were concluded to be 50mg/kg/day in rats.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 98%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 99%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, ≥98%