Merck
Fragrance material review on benzyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-24)
A Lapczynski, D McGinty, L Jones, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of benzyl salicylate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Benzyl salicylate, purum, ≥99.0% (GC)
Supelco
Benzyl salicylate, analytical standard