Merck
  • Regiospecific solid-phase strategy to N7-substituted purines and its application to 8-azapurines and [I]-condensed purines.
Regiospecific solid-phase strategy to N7-substituted purines and its application to 8-azapurines and [I]-condensed purines.
Journal of combinatorial chemistry (2007-08-11)
Han Fu, Mark Kheng Chuan Tan, Yulin Lam
ABSTRACT
A highly regioselective and traceless solid-phase route to 1,7,8-trisubstituted purines has been developed. This methodology could be extended to the preparation of 8-azapurines and [i]-condensed purines. A representative set of 17 purines, azapurines, and [i]-condensed purines was synthesized. This paper also describes a mild method to prepare the p-benzyloxybenzylamine (BOBA) resin.