Merck
  • [Putative mycotoxin-producing fungi isolated from alpataco (Prosopis flexuosa) fruits].
[Putative mycotoxin-producing fungi isolated from alpataco (Prosopis flexuosa) fruits].
Revista iberoamericana de micologia (2007-06-27)
Mónica Canafoglia, Ricardo Comerio, Virginia Fernández Pinto, Graciela Vaamonde
ABSTRACT
Fungi contaminant of alpataco (Prosopis flexuosa) fruits from La Pampa province (Argentina) were identified. Alternaria alternata and Sphaeropsis sapinea were the dominant species. Phoma sp., Nigrospora sp., Preussia minima, Cladosporium sp., Pithomyces chartarum, Epicoccum nigrum, Aspergillus niger and Aspergillus speluneus were also isolated but with less frequency. Twelve strains of Alternaria alternata, the toxigenic species with higher incidence, were screened for alternariol (AOH), alternariol monomethyl ether (AME) and tenuazonic acid (TA) production. Since one isolate was able to produce AME, six isolates produced AOH and AME and two isolates produced AOH, AME and TA, these results indicate a potential risk of contamination with Alternaria toxins in this substrate.
Sigma-Aldrich
Tenuazonic acid copper salt from Alternaria alternata
Supelco
Tenuazonic acid, analytical standard