- A novel compound, depudecin, induces production of transformation to the flat phenotype of NIH3T3 cells transformed by ras-oncogene.
A novel compound, depudecin, induces production of transformation to the flat phenotype of NIH3T3 cells transformed by ras-oncogene.
Biochemical and biophysical research communications (1992-01-15)
K Sugita, H Yoshida, M Matsumoto, S Matsutani
PMID1731795
ABSTRACT
A novel compound, depudecin, induced production of the flat phenotype of Ki-ras-transformed NIH3T3 cells at the low concentration of 1 microgram/ml. This effect was reversible. Actin stress fiber was detected in these cells after depudecin treatment. Almost complete reversion to the flat phenotype was observed at 6 h after depudecin addition. The synthesis of ras-mRNA did not decrease enough with depudecin treatment at the concentration of 10 micrograms/ml to reverse the transformed morphology.