Merck
Solid-state phase transition induced by pressure in LiOH x H2O.
The journal of physical chemistry. B (2006-07-11)
Elisa Di Pietro, Marco Pagliai, Gianni Cardini, Vincenzo Schettino
ABSTRACT
When the free energy surface of the lithium hydroxide monohydrate crystal was explored, the high-pressure solid-state phase transition was determined. The high-pressure phase has been obtained through ab initio Car-Parrinello molecular dynamics simulation in the isothermic-isobaric ensemble. The recent metadynamics method has been applied to overcome the high activation energy barriers typical of rare events, like solid-state phase transition at high pressures. In the LiOH x H2O system, there are two kinds of H bonds: water-water and hydroxyl-water. The effect of the pressure has been investigated, to give further insight into the high-pressure phase. The strengthening of the H bonds of the system produces modifications in the water and the hydroxyl ion dipole electronic environment. The infrared spectra of both phases have been calculated and compared with experiments, and the assignment of the external modes has been discussed.
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%