Merck
  • Micronucleus studies in the peripheral blood and bone marrow of mice treated with jet fuels, JP-8 and Jet-A.
Micronucleus studies in the peripheral blood and bone marrow of mice treated with jet fuels, JP-8 and Jet-A.
Mutation research (2006-07-04)
Vijayalaxmi, Andrew D Kligerman, Thomas J Prihoda, Stephen E Ullrich
ABSTRACT
The potential adverse effects of dermal and inhalation exposure of jet fuels are important for health hazard evaluation in humans. The genotoxic potential of jet fuels, JP-8 and Jet-A, was investigated in an animal model. Mice were treated dermally with either a single or multiple applications of these jet fuels. Peripheral blood and bone marrow smears were prepared to examine the incidence of micronuclei (MN) in polychromatic erythrocytes (PCEs). In all experiments, using several different exposure regimens, no statistically significant increase in the incidence of MN was observed in the bone marrow and/or peripheral blood of mice treated with JP-8 or Jet-A when compared with those of untreated control animals. The data in mice treated with a single dose of JP-8 or Jet-A did not confirm the small but statistically significant increase in micronuclei reported in our previous study.
Sigma-Aldrich
Kerosene, SAJ special grade, ≥90.0%
Sigma-Aldrich
Kerosene, CP
Sigma-Aldrich
Kerosene, purum
Sigma-Aldrich
Kerosene, reagent grade, low odor