Merck
  • Monosaccharides as internal probes for the determination of the absolute configuration of 2-butanol.
Monosaccharides as internal probes for the determination of the absolute configuration of 2-butanol.
Magnetic resonance in chemistry : MRC (2005-12-17)
Patrycja Seroka, Marcin Płosiński, Jacek Czub, Paweł Sowiński, Jan Pawlak
ABSTRACT
D-Glucose, D-mannose and L-rhamnose were reacted with a racemic mixture of 2-butanol, and the resulting alpha-glycosides were analyzed by 1H NMR with COSY and NOESY experiments. Conformational analysis of alpha-glycosidic bonds performed with molecular modeling and appropriate heteronuclear long-range coupling measurements and combined with analysis of dipolar couplings observed in NOESY spectra allowed the assignment of absolute configuration in the aglycones of elucidated alpha-glycosides.
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%
Sigma-Aldrich
2-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Aluminum-tri-sec-butoxide, 97%
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Supelco
2-Butanol, analytical standard
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%