Merck
  • Mother rotors and the mechanisms of D600-induced type 2 ventricular fibrillation.
Mother rotors and the mechanisms of D600-induced type 2 ventricular fibrillation.
Circulation (2004-10-07)
Tsu-Juey Wu, Shien-Fong Lin, Ali Baher, Zhilin Qu, Alan Garfinkel, James N Weiss, Chih-Tai Ting, Peng-Sheng Chen
ABSTRACT
Two types of ventricular fibrillation (VF) have been demonstrated in isolated rabbit hearts during D600 infusion. Type 1 VF is characterized by the presence of multiple, wandering wavelets, whereas type 2 VF shows local spatiotemporal periodicity. We hypothesized that a single mother rotor underlies type 2 VF. One (protocol I) or 2 (protocol II) cameras were used to map the epicardial ventricular activations in Langendorff-perfused rabbit hearts. Multiple episodes of type 2 VF were induced in 22 hearts by high-concentration (> or =2.5 mg/L) D600 (protocol I). During type 2 VF, a single spiral wave (n=19) and/or an epicardial breakthrough pattern (n=11) was present in 14 hearts. These spiral waves either slowly drifted or intermittently anchored on the papillary muscle (PM) of the left ventricle. Dominant-frequency (DF) analyses showed that the highest local DF was near the PM (12.5+/-1.1 Hz). There was an excellent correlation between the highest local DF of these spiral waves and breakthroughs (11.8+/-1.7 Hz) and the DF of simultaneously obtained global pseudo-ECG (11.2+/-1.8 Hz, r=0.97, P<0.0001) during type 2 VF. We also successfully reproduced the major features of type 2 VF by using the Luo-Rudy action-potential model in a simulated, 3-dimensional tissue slab, under conditions of reduced excitability and flat action-potential duration restitution. Either a stationary or a slowly drifting mother rotor can result in type 2 VF. Colocalization of the stationary mother rotors with the PM suggests the importance of underlying anatomic structures in mother rotor formation.
Sigma-Aldrich
(±)-Methoxyverapamil hydrochloride, ≥98%