Merck
  • Aerobic oxidation of thiols to disulfides catalyzed by trichlorooxyvanadium.
Aerobic oxidation of thiols to disulfides catalyzed by trichlorooxyvanadium.
Chemical & pharmaceutical bulletin (2004-05-11)
Masayuki Kirihara, Kumiko Okubo, Tomoyuki Uchiyama, Yoko Kato, Yuta Ochiai, Shinya Matsushita, Akihiko Hatano, Kan Kanamori
ABSTRACT
Thiols were converted into disulfide by the aerobic oxidation catalyzed by trichlorooxyvanadium in the presence of molecular sieves 3A.
Sigma-Aldrich
Vanadium(V) oxychloride, 99%