Merck
  • Spectrophotometric study of the system Hg(II)-thymol blue-H2O and its evidence through electrochemical means.
Spectrophotometric study of the system Hg(II)-thymol blue-H2O and its evidence through electrochemical means.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2004-01-30)
P Balderas-Hernández, A Rojas-Hernández, M Galván, M T Ramírez-Silva
ABSTRACT
The detailed analysis of the experimental spectrophotometric data obtained from solutions containing the acid-base indicator thymol blue (TB) and mercury(II) (Hg(II)) coupled with data processing by means of the SQUAD program, a chemical model was determined that includes the formation of complexes indicator-metal ion (HgTB and HgOTB), dimer species (H3TB2 and H4TB2) and monomer species (HTB and TB). The values of the overall formation constants (log beta) were calculated for the chemical equilibria involved: TB+Hg<-->HgTB log beta=16.047 +/- 0.043, TB+Hg+H2O<-->HgOHTB+H log beta=7.659 +/- 0.049, 2TB+4H<-->H4TB2 log beta=31.398 +/- 0.083, 2TB+3H<-->H3TB2 log beta=29.953 +/- 0.084 and H+TB<-->HTB-log beta=8.900. To compliment the present research, the values of the absorptivity coefficients are included for all the species involved, within a wide range of wavelengths (250-700 nm). The latter were used subsequently to carry simulations of the absorption spectra at various pH values, thus corroborating that the chemical model proposed is fully capable to describe the experimental information. Voltammetric study performed evidenced the formation of a complex with a 1:1 stoichiometry Hg(II):TB.
Sigma-Aldrich
Thymol Blue, JIS special grade
Sigma-Aldrich
Thymolphthalein, JIS special grade
Sigma-Aldrich
Thymol Blue, ACS reagent
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymol Blue sodium salt, ACS reagent, Dye content 95 %
Supelco
Thymolphthalein, ACS reagent, indicator (pH 8.8-10.5)