Merck
Silver carbonate staining reveals mitochondrial heterogeneity.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2004-01-20)
José M López-Cepero
ABSTRACT
Silver staining methods, when selective, yield a high-contrast and high-resolution image in optical microscopy. A classical method for silver impregnation of mitochondria has been applied to murine tissues and reveals a marked heterogeneity among mitochondria in single cells. This heterogeneity can be detected in the optical microscope but is even more evident at the ultrastructural level. The differences in staining intensity may reflect different stages in the mitochondrial life cycle. The progressive accumulation of uranyl-argyrophilic material may be a marker of mitochondrial aging. This highly selective staining procedure may be of use in studies of mitochondrial changes under pathological conditions and during apoptosis.
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver carbonate, 99%
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading