Merck
  • Blood pharmacokinetics of tertiary amyl methyl ether in male and female F344 rats and CD-1 mice after nose-only inhalation exposure.
Blood pharmacokinetics of tertiary amyl methyl ether in male and female F344 rats and CD-1 mice after nose-only inhalation exposure.
Journal of applied toxicology : JAT (2003-11-25)
Susan C J Sumner, Derek B Janszen, Bahman Asgharian, Timothy A Moore, Carol M Bobbitt, Timothy R Fennell
ABSTRACT
Interest in understanding the biological behavior of aliphatic ethers has increased owing to their use as gasoline additives. The purpose of this study was to investigate the blood pharmacokinetics of the oxygenate tertiary amyl methyl ether (TAME), its major metabolite tertiary amyl alcohol (TAA) and acetone in rats and mice following inhalation exposure to TAME. Species differences in the area under the curve (AUC) for TAME were significant at each exposure concentration. For rats, the blood TAME AUC increased in proportion with an increase in exposure concentration. For mice, an increase in exposure concentration (100-500 ppm) resulted in a disproportional increase in the TAME AUC. Mice had greater (two- to threefold) blood concentrations of TAA compared with rats following exposure to 2500 or 500 ppm TAME. Mice had a disproportional increase in the TAA AUC with an increase in exposure concentration (100-500 ppm). This difference could result from saturation of a process (e.g. oxidation, glucuronide conjugation) that is involved in the further metabolism of TAA. For each species, gender and exposure concentration, acetone increased during exposure and returned to control values by 16 h following exposure. The source of acetone could be both as a metabolite of TAA or an effect on endogenous metabolism produced by exposure to TAME.
Product Description
Supelco
tert-Amyl methyl ether, analytical standard
Sigma-Aldrich
tert-Amyl methyl ether, 97%