- Identification, isolation, and structural studies of the outer membrane lipopolysaccharide of Caulobacter crescentus.
Identification, isolation, and structural studies of the outer membrane lipopolysaccharide of Caulobacter crescentus.
The lipopolysaccharide (LPS) of the outer membrane of Caulobacter crescentus was purified and analyzed. Two distinct strains of the species, NA 1000 and CB2A, were examined; despite differences in other membrane-related polysaccharides, the two gave similar LPS composition profiles. The LPS was the equivalent of the rough LPS described for other bacteria in that it lacked the ladder of polysaccharide-containing species that results from addition of variable amounts of a repeated sequence of sugars, as detected by gel electrophoresis in smooth LPS strains. The purified LPS contained two definable regions: (i) an oligosaccharide region, consisting of an inner core of three residues of 2-keto-3-deoxyoctonate, two residues of alpha-L-glycero-D-mannoheptose, and one alpha-D-glycero-D-mannoheptose unit and an outer core region containing one residue each of alpha-D-mannose, alpha-D-galactose, and alpha-D-glucose, with the glucose likely phosphorylated and (ii) a region equivalent to the lipid A region of the archetype, consisting primarily of an esterified fatty acid, 3-OH-dodecanoate. The lipid A-like region was resistant to conclusive analysis; in particular, although a variety of analytical methods were used, no amino sugars were detected, as is found in the lipid A of the LPS of most bacteria.