Merck
Phosphorous pentachloride chemical burn--a slowly healing injury.
Burns : journal of the International Society for Burn Injuries (1992-08-01)
A Eldad, M Chaouat, A Weinberg, A Neuman, P Ben Meir, M Rotem, M R Wexler
ABSTRACT
A 51-year-old chemical engineer sustained phosphorous pentachloride partial skin thickness burns over 20 per cent of his body surface area. Although macroscopically and microscopically the wound seemed to be superficial, the course of clinical healing of this injury was very slow and painful. Retrospectively this burn should have been treated by early excision and grafting.
Sigma-Aldrich
Phosphorus trichloride, 99.999% trace metals basis
Sigma-Aldrich
Phosphorus trichloride, ReagentPlus®, 99%