Merck
  • Relationship between chromatographic properties of aromatic carboxylic acids and their structure.
Relationship between chromatographic properties of aromatic carboxylic acids and their structure.
Journal of chromatography. A (2003-07-02)
Olga Grechishkina
ABSTRACT
The retention of aromatic carboxylic acids and their esters in reversed-phase HPLC is proportional to the sum of their partition coefficients in octanol-water system and retention increments. This equation can be used for the identification of various side-products formed in monomer production.
Sigma-Aldrich
1,2,4,5-Benzenetetracarboxylic acid, 96%
Sigma-Aldrich
p-Toluic acid, 98%