Merck
  • A dynamically modified microfluidic poly(dimethylsiloxane) chip with electrochemical detection for biological analysis.
A dynamically modified microfluidic poly(dimethylsiloxane) chip with electrochemical detection for biological analysis.
Electrophoresis (2002-11-02)
Yue-Hua Dou, Ning Bao, Jing-Juan Xu, Hong-Yuan Chen
ABSTRACT
Separation and direct detection of amino acids, glucose and peptide in a 3.1 cm separation channel made of poly(dimethylsiloxane) (PDMS) with end-column amperometric detection at a copper microdisk electrode was developed. This system is the integration of a normal sized working electrode with electrochemical detection on a PDMS microfabricated device. The PDMS channels dynamically modified by 2-morpholinoethanesulfonic acid (MES) show less adsorption and more enhanced efficiency than that of unmodified ones when applied to separations of these biological molecules. The migration time is less than 100 s and the reproducibility of migration time is satisfactory with relative standard deviation (RSD) of 2.8% in 19 successive injections. The limits of detection of arginine (Arg), glucose, and methionine-glycine (Met-Gly) are estimated to be 2.0, 8.5, and 64.0 microM at S/N = 3, approximately 0.5-16.0 fmol, respectively. Variances influencing the separation efficiency and amperometric response, including injection, separation voltage, detection potential, or concentration of buffer and additive, are assessed and optimized.
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
SAFC
MES sodium salt
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Met-Gly
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)