Merck
  • Asymmetric synthesis of 2-substituted piperidines using a multi-component coupling reaction: rapid assembly of (S)-coniine from (S)-1-(1-phenylethyl)-2-methyleneaziridine.
Asymmetric synthesis of 2-substituted piperidines using a multi-component coupling reaction: rapid assembly of (S)-coniine from (S)-1-(1-phenylethyl)-2-methyleneaziridine.
Chemical communications (Cambridge, England) (2002-09-21)
J F Hayes, M Shipman, H Twin
ABSTRACT
(S)-Coniine is made using a reaction which assembles the piperidine ring by the sequential formation of four new chemical bonds and installs the C-2 stereogenic centre with high levels of diastereocontrol (90% de).