Merck
An assessment of "Caloreen", a new, high-energy source.
Medical journal of Zambia (1978-10-01)
T K Lambart, L Worsonu
PMID121187
ABSTRACT
A prospective evaluation study has been undertaken on "Caloreen", a new high energy preparation. Data were complete in 23 consecutive patients. Mean weight gain was 2.6 Kg (adults), and 2.2 Kg (children). Levels of haemoglobin and of serum albumin were. maintained (10.9-10.9 g%, and 3.4-3.6 g% respectively). Patient compliance was high. It was concluded that "Caloreen" was useful, and would given even more satisfactory results in predominantly Paediatric practices.
Sigma-Aldrich
Dextrin from corn, Type II, powder
Millipore
Dextrin from potato starch, suitable for microbiology
Sigma-Aldrich
Dextrin from corn, Type I, powder