Merck
  • Mononuclear platinum(II) complex with 2-phenylpyridine ligands showing high cytotoxicity against mouse sarcoma 180 cells acquiring high cisplatin resistance.
Mononuclear platinum(II) complex with 2-phenylpyridine ligands showing high cytotoxicity against mouse sarcoma 180 cells acquiring high cisplatin resistance.
Journal of medicinal chemistry (2001-12-14)
T Okada, I M El-Mehasseb, M Kodaka, T Tomohiro, K Okamoto, H Okuno
ABSTRACT
Mouse sarcoma 180 cell with a 25-fold higher cisplatin (CDDP) resistance, termed S-180cisR, is newly established. S-180cisR cells grow quite slowly in the presence of CDDP with high concentration. This may show that S-180cisR cells modulate the cell cycle to acquire CDDP resistance. P-Glycoprotein is selectively expressed on the surface of S-180cisR, which is not on CDDP-sensitive S-180 parent cells. In an experiment using an inhibitor (verapamil) of P-glycoprotein, cytotoxicity of CDDP against S-180cisR is significantly increased (viz., IC(50) value is decreased) and accumulation of CDDP in S-180cisR cells is also increased. These results indicate that enhanced pumping-out of CDDP by P-glycoprotein should be one of the reasons for the CDDP resistance of S-180cisR. A platinum(II) complex with a cyclometalated 2-phenylpyridine ligand and a nonchelated one (complex 5) is synthesized, and its structure is determined by X-ray structural analysis. Complex 5 has a cyctotoxicity against S-180cisR higher than that of CDDP and its derivatives with 2- or 3-substituted pyridine ligands (complexes 2-4, 6, 7). Complex 5 is incorporated in S-180cisR to an enormously greater extent than CDDP; that is, the ratio of accumulated platinum amount after 3 h is 61.9. In S-180 parent cells, on the other hand, the ratio remains 8.1. This high accumulation of complex 5 into S-180cisR must account for the higher activity of complex 5 against S-180cisR compared to CDDP.
Sigma-Aldrich
2-Phenylpyridine, 98%