Merck
Enzymatic synthesis of aza-l-tyrosines.
Bioorganic & medicinal chemistry letters (2001-08-22)
E B Watkins, R S Phillips
ABSTRACT
Tyrosine phenol-lyase from Citrobacter freundii synthesizes 2-aza-L-tyrosine and 3-aza-L-tyrosine from 3-hydroxypyridine and 2-hydroxypyridine, respectively, and ammonium pyruvate.
Sigma-Aldrich
3-Hydroxypyridine, 98%