Merck
Anthelmintic resistance on an organized sheep farm in India.
Tropical animal health and production (2001-07-28)
C P Swarnkar, P K Sanyal, D Singh, F A Khan, P S Bhagwan
ABSTRACT
Monitoring anthelmintic resistance in strongyle nematodes by the faecal egg count reduction test and a commercial larval development assay on an organized sheep farm in the semi-arid area of Rajasthan revealed the emergence of resistance to benzimidazoles and rafoxanide and a potential risk of the development of levamisole/tetramisole resistance. A benzimidazole/levamisole combination, avermectins and closantel were each found to be efficacious.
Product Description
Supelco
Rafoxanide, PESTANAL®, analytical standard
Pricing and availability is not currently available.