Merck
Total synthesis of reveromycin A.
Organic letters (2000-07-13)
T Shimizu, T Masuda, K Hiramoto, T Nakata
ABSTRACT
The stereoselective total synthesis of reveromycin A (1), a potent inhibitor of eukaryotic cell growth, has been accomplished on the basis of the stereocontrolled construction of the 6,6-spiroketal system, efficient succinylation of the tert-alcohol under high pressure, and the introduction of the unsaturated side chains.
Sigma-Aldrich
Reveromycin A, solid