- Solar photocatalytic mineralization of commercial pesticides: acrinathrin.
Solar photocatalytic mineralization of commercial pesticides: acrinathrin.
A comparative study of the degradation of commercial acrinathrin spiked in water using TiO2 photocatalysis and photolysis under sunlight was performed. Samples were analysed by liquid chromatography-diode array detector (HPLC-DAD) and gas chromatography-ion trap-mass spectrometric detector (GC-ITMS). Additional total organic carbon (TOC) analyses were carried out to evaluate the mineralisation rates. One photoproduct, 2-phenoxy benzaldehyde, was unequivocally identified and evaluated by GC-ITMS during the processes. Although acrinathrin is almost destroyed when exposed to irradiation for more than 400 h, photocatalysis with TiO2 noticeably reduced degradation to a few hours. In this case, with the additional presence of peroxydisulphate, in less than 2 h acrinathrin is completely destroyed. Mineralisation of acrinathrin, without catalyst, was only around 50% after 400 h of irradiation.