Merck
  • [An experimental study on effects of antimony on anti-oxidase system on liver mitochondria in mice].
[An experimental study on effects of antimony on anti-oxidase system on liver mitochondria in mice].
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine] (1999-05-14)
A Wang, F An, Z Gao
ABSTRACT
To explore the mechanisms of toxic liver damage caused by antimony. Experimental mice were injected peritoneally 40 mg/kg of antimony trioxide daily for 28 days to observe the effects of it on the anti-oxidase system in their liver mitochondria. Activities of aspartate aminotransferase and alanine aminotransferase increased obviously in the experimental animals, and those of superoxide dismutase and glutathione peroxidase in liver mitochondria lowered, as compared to those in the control animals with statistical significance. Content of malonyl dialdehyde in the liver mitochondria was 47.27 +/- 1.21 mumol/L in the experimental animals, significantly higher than that in controls. It suggested that the mechanism of liver damage caused by antimony associated with injury to the ability of anti-oxidation and lipid peroxidation in the liver mitochondria.
Sigma-Aldrich
Antimony(III) chloride, JIS special grade, ≥98.0%
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Antimony(III) chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
Antimony(III) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Antimony standard solution, 1000 ppm Sb, suitable for atomic absorption spectrometry