Merck
POLG mutations and Alpers syndrome.
Annals of neurology (2005-06-02)
Guido Davidzon, Michelangelo Mancuso, Silvio Ferraris, Catarina Quinzii, Michio Hirano, Heidi L Peters, Denise Kirby, David R Thorburn, Salvatore DiMauro
ABSTRACT
Alpers-Huttenlocher syndrome (AHS) an autosomal recessive hepatocerebral syndrome of early onset, has been associated with mitochondrial DNA (mtDNA) depletion and mutations in polymerase gamma gene (POLG). We have identified POLG mutations in four patients with hepatocerebral syndrome and mtDNA depletion in liver, who fulfilled criteria for AHS. All were compound heterozygous for the G848S and W748S mutations, previously reported in patients with progressive external ophtalmoplegia or ataxia. We conclude that AHS should be included in the clinical spectrum of mtDNA depletion and is often associated with POLG mutations, which can cause either multiple mtDNA deletions or mtDNA depletion.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2