Merck
Overturning indolyne regioselectivities and synthesis of indolactam V.
Journal of the American Chemical Society (2011-03-01)
Sarah M Bronner, Adam E Goetz, Neil K Garg
ABSTRACT
We report the design and synthesis of an indolyne that displays a reversal in regioselectivity, in both nucleophilic addition and cycloaddition reactions, compared to typical 4,5-indolynes. Our approach utilizes simple computations to predict regioselectivity in reactions of unsymmetrical arynes. With this methodology, novel benzenoid-substituted indoles can be accessed with significant regiocontrol. Furthermore, the technology provides an unconventional tactic for the synthesis of C4-substituted indole alkaloids, as demonstrated by a synthesis of indolactam V.
Sigma-Aldrich
(−)-Indolactam V, ≥96% (HPLC)