Merck
  • Wortmannin inhibits translation of tumor necrosis factor-alpha in superantigen-activated T cells.
Wortmannin inhibits translation of tumor necrosis factor-alpha in superantigen-activated T cells.
International immunology (1999-08-28)
M Ramírez, N Fernández-Troy, M Buxadé, R P Casaroli-Marano, D Benítez, C Pérez-Maldonado, E Espel
ABSTRACT
The superantigen toxic shock syndrome toxin (TSST)-1 can induce tumor necrosis factor (TNF)-alpha expression in T cells and monocytes, through different signaling pathways. We have stimulated peripheral blood mononuclear cells with TSST-1 and found that the major cell producers of TNF-alpha as detected by cytofluorimetry and immunocytochemistry were CD4(+) T lymphocytes. The expression of TNF-alpha by CD4(+) T cells can be inhibited by either, wortmannin (WN) or LY 294002, two phosphatidylinositol 3-kinase (PI 3-K) inhibitors. The inhibitory effect is not transcriptional as WN does not change the mRNA steady state of TNF-alpha at any of the concentrations tested and LY 294002 when preincubated with mononuclear cells at its median inhibitory concentration (IC(50) = 1. 4 microM) significantly inhibited the expression of TNF-alpha but not its mRNA. Immunoprecipitation of pulse-labeled intracellular TNF-alpha showed a specific decrease in the synthesis of this cytokine on cells treated with PI 3-K inhibitors. The p38 mitogen-activated protein kinase (MAPK) is involved in control of TNF-alpha translation in human macrophages. In T cells, we have found that the p38 MAPK inhibitor SB 203580 significantly decreased the secretion of TNF-alpha but not its mRNA. In addition, the combined use of WN and SB 203580 had an additive inhibitory effect on secretion of TNF-alpha. Therefore, both PI 3-K and p38 MAPK signaling pathways control TNF-alpha production in T cells.