Merck
  • Inhibition of heat-induced denaturation of albumin by nonsteroidal antiinflammatory drugs (NSAIDs): pharmacological implications.
Inhibition of heat-induced denaturation of albumin by nonsteroidal antiinflammatory drugs (NSAIDs): pharmacological implications.
Archives of pharmacal research (2001-05-08)
L Saso, G Valentini, M L Casini, E Grippa, M T Gatto, M G Leone, B Silvestrini
ABSTRACT
The activity of nonsteroidal antiinflammatory drugs (NSAIDs) in rheumatoid arthritis is not only due to the inhibition of the production of prostaglandins, which can even have beneficial immunosuppressive effects in chronic inflammatory processes. Since we speculated that these drugs could also act by protecting endogenous proteins against denaturation, we evaluated their effect on heat-induced denaturation human serum albumin (HSA) in comparison with several fatty acids which are known to be potent stabilizers of this protein. By the Mizushimas assay and a recently developed HPLC assay, we observed that NSAIDs were slightly less active [EC50 to approximately 10(-5)-10(-4) M] than FA and that the HPLC method was less sensitive but more selective than the turbidimetric assay, i.e. it was capable of distinguishing true antiaggregant agents like FA and NSAIDs from substances capable of inhibiting the precipitation of denatured protein aggregates. In conclusion, this survey could be useful for the development of more effective agents in protein condensation diseases like rheumatic disorders, cataract and Alzheimers disease.
Sigma-Aldrich
Sulindac, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Meclofenamic acid sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Pricing and availability is not currently available.