Merck
  • Association of the I148M/PNPLA3 variant with elevated alanine transaminase levels in normal-weight and overweight/obese Mexican children.
Association of the I148M/PNPLA3 variant with elevated alanine transaminase levels in normal-weight and overweight/obese Mexican children.
Gene (2013-03-21)
Elena Larrieta-Carrasco, Paola León-Mimila, Teresa Villarreal-Molina, Hugo Villamil-Ramírez, Sandra Romero-Hidalgo, Leonor Jacobo-Albavera, Roxana Gutiérrez-Vidal, Blanca E López-Contreras, Luz E Guillén-Pineda, Fausto Sánchez-Muñoz, Rafael Bojalil, Ana M Mejía-Domínguez, Nahúm Méndez-Sánchez, Aaron Domínguez-López, Carlos A Aguilar-Salinas, Samuel Canizales-Quinteros
ABSTRACT
Non-alcoholic fatty liver disease (NAFLD) and elevated alanine transaminase (ALT) levels are common in obese Hispanic adults and children. Recently, a PNPLA3 gene variant (I148M) was strongly associated with NAFLD and higher ALT levels in obese adults, including Hispanics. The aims of this study were to estimate the frequency of elevated ALT levels, and to address the influence of obesity and PNPLA3/I148M on ALT levels in a general population sample of Mexican school-aged children. A total of 1037 non-related Mexican children aged 6 to 12 years were genotyped for the I148M variant. Anthropometric, clinical and metabolic parameters were collected from all participants. Elevated ALT levels (>35 U/L) were more frequent in obese (26.9%) and overweight (9.3%) than in normal weight children (2.2%). The M148M genotype was significantly associated with elevated ALT levels in this population (OR=3.7, 95% CI 2.3-5.9; P=3.7×10(-8)), and children carrying the M148M genotype showed significantly lower HDL cholesterol levels and BMI z-core (P=0.036 and 0.015, respectively). On stratifying by BMI percentile, this genotype conferred a much greater risk of elevated ALT levels in normal weight (OR=19.9, 95% CI 2.5-157.7; P=0.005) than overweight and obese children (OR=3.4, 95% CI 1.3-8.9; P=0.014 and OR=3.1, 95% CI 1.7-5.5; P=1.4 x10(-4), respectively). The I148M PNPLA3 variant is strongly associated with elevated ALT levels in normal weight and overweight/obese Mexican children. Thus, the M148M genotype may be considered as an important risk factor for liver damage in this population.
Product Description
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
SAFC
L-Isoleucine
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid