Merck
  • Immunoendocrine responses of male spinal cord injured athletes to 1-hour self-paced exercise: pilot study.
Immunoendocrine responses of male spinal cord injured athletes to 1-hour self-paced exercise: pilot study.
Journal of rehabilitation research and development (2013-01-10)
Judith E Allgrove, Mark Chapman, Tatiana Christides, Paul M Smith
ABSTRACT
This study examined the effect of a 1 h, self-paced handcycling time trial on blood leukocytes, mucosal immunity, and markers of stress in paraplegic athletes. Nine male paraplegic athletes (spinal injury level thoracic 4-lumbar 2) performed 1 h of handcycling exercise on a standard 400 m athletics track. Heart rate (HR) was measured continuously during exercise, and a retrospective rating of perceived exertion (RPE) was obtained immediately after. Venous blood and saliva samples were collected immediately before exercise (Pre-Ex), after exercise (End-Ex), and 1 h postexercise (1-h Post). The athletes completed mean +/- standard error of mean 22.4 +/- 1.1 km cycling at HR 165 +/- 2 beats/min, RPE 15 +/- 1, and blood lactate 7.9 +/- 2.5 mmol/L. Total leukocytes increased 72% and neutrophils increased 74% End-Ex; both remained elevated at 1-h Post (both p < 0.05). Lymphocytes increased 53% and natural killer cells increased 175% End-Ex (both p < 0.05), but returned to near baseline levels 1-h Post. Increases (p < 0.05) were observed End-Ex in alpha-amylase activity (p < 0.05), which returned to baseline at 1-h Post, but there was no significant change in saliva flow rate, salivary immunoglobulin A, or cortisol. These data confirm that 1 h of handcycling exercise elevated circulating leukocytes but had a minimal effect on mucosal immunity. These changes appear to be associated with alpha-amylase rather than cortisol.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)