Merck
  • TRANCE is a novel ligand of the tumor necrosis factor receptor family that activates c-Jun N-terminal kinase in T cells.
TRANCE is a novel ligand of the tumor necrosis factor receptor family that activates c-Jun N-terminal kinase in T cells.
The Journal of biological chemistry (1997-10-06)
B R Wong, J Rho, J Arron, E Robinson, J Orlinick, M Chao, S Kalachikov, E Cayani, F S Bartlett, W N Frankel, S Y Lee, Y Choi
ABSTRACT
A novel member of the tumor necrosis factor (TNF) cytokine family, designated TRANCE, was cloned during a search for apoptosis-regulatory genes using a somatic cell genetic approach in T cell hybridomas. The TRANCE gene encodes a type II membrane protein of 316 amino acids with a predicted molecular mass of 35 kDa. Its extracellular domain is most closely related to TRAIL, FasL, and TNF. TRANCE is an immediate early gene up-regulated by TCR stimulation and is controlled by calcineurin-regulated transcription factors. TRANCE is most highly expressed in thymus and lymph nodes but not in nonlymphoid tissues and is abundantly expressed in T cells but not in B cells. Cross-hybridization of the mouse cDNA to a human thymus library yielded the human homolog, which encodes a protein 83% identical to the mouse ectodomain. Human TRANCE was mapped to chromosome 13q14 while mouse TRANCE was located to the portion of mouse chromosome 14 syntenic with human chromosome 13q14. A recombinant soluble form of TRANCE composed of the entire ectodomain induced c-Jun N-terminal kinase (JNK) activation in T cells but not in splenic B cells or in bone marrow-derived dendritic cells. These results suggest a role for this TNF-related ligand in the regulation of the T cell-dependent immune response.