Merck
  • Divergent and Modular Synthesis of Terpenoid Scaffolds via a AuI Catalyzed One-Pot Cascade.
Divergent and Modular Synthesis of Terpenoid Scaffolds via a AuI Catalyzed One-Pot Cascade.
Angewandte Chemie (International ed. in English) (2021-10-30)
Huy Tran, Guillaume Revol, Alyson Poyser, Louis Barriault
ABSTRACT
A one-pot cascade sequence to generate synthetically challenging polycyclic scaffolds is reported utilizing a novel Lewis acid gold catalyst for the key cyclization step, enabling the divergent synthesis of both 6,6,5-tricyclic and 6,6,6,5-tetracyclic cores through both ligand and reaction condition control. We have combined the intrinsic complexity and stereoselectivity of cycloadditions with the electronic and steric properties of gold complexes to selectively generate complex polycyclic scaffolds in a single operation.
Sigma-Aldrich
Me4tButylXphos, 96%