Merck
  • Auto-antibodies against type I IFNs in > 10% of critically ill COVID-19 patients: a prospective multicentre study.
Auto-antibodies against type I IFNs in > 10% of critically ill COVID-19 patients: a prospective multicentre study.
Annals of intensive care (2023-01-01)
Romain Arrestier, Paul Bastard, Thibaut Belmondo, Guillaume Voiriot, Tomas Urbina, Charles-Edouard Luyt, Adrian Gervais, Lucy Bizien, Lauriane Segaux, Mariem Ben Ahmed, Raphaël Bellaïche, Taï Pham, Zakaria Ait-Hamou, Damien Roux, Raphael Clere-Jehl, Elie Azoulay, Stéphane Gaudry, Julien Mayaux, Nicolas Fage, Hafid Ait-Oufella, Elsa Moncomble, Mélodie Parfait, Karim Dorgham, Guy Gorochov, Armand Mekontso-Dessap, Florence Canoui-Poitrine, Jean-Laurent Casanova, Sophie Hue, Nicolas de Prost
ABSTRACT
Auto-antibodies (auto-Abs) neutralizing type I interferons (IFN) have been found in about 15% of critical cases COVID-19 pneumonia and less than 1% of mild or asymptomatic cases. Determining whether auto-Abs influence presentation and outcome of critically ill COVID-19 patients could lead to specific therapeutic interventions. Our objectives were to compare the severity at admission and the mortality of patients hospitalized for critical COVID-19 in ICU with versus without auto-Abs. We conducted a prospective multicentre cohort study including patients admitted in 11 intensive care units (ICUs) from Great Paris area hospitals with proven SARS-CoV-2 infection and acute respiratory failure. 925 critically ill COVID-19 patients were included. Auto-Abs neutralizing type I IFN-α2, β and/or ω were found in 96 patients (10.3%). Demographics and comorbidities did not differ between patients with versus without auto-Abs. At ICU admission, Auto-Abs positive patients required a higher FiO2 (100% (70-100) vs. 90% (60-100), p = 0.01), but were not different in other characteristics. Mortality at day 28 was not different between patients with and without auto-Abs (18.7 vs. 23.7%, p = 0.279). In multivariable analysis, 28-day mortality was associated with age (adjusted odds ratio (aOR) = 1.06 [1.04-1.08], p < 0.001), SOFA score (aOR = 1.18 [1.12-1.23], p < 0.001) and immunosuppression (aOR = 1.82 [1.1-3.0], p = 0.02), but not with the presence of auto-Abs (aOR = 0.69 [0.38-1.26], p = 0.23). In ICU patients, auto-Abs against type I IFNs were found in at least 10% of patients with critical COVID-19 pneumonia. They were not associated with day 28 mortality.
Sigma-Aldrich
IFN-omega human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture