Merck
  • Isotropic three-dimensional dual-color super-resolution microscopy with metal-induced energy transfer.
Isotropic three-dimensional dual-color super-resolution microscopy with metal-induced energy transfer.
Science advances (2022-06-09)
Jan Christoph Thiele, Marvin Jungblut, Dominic A Helmerich, Roman Tsukanov, Anna Chizhik, Alexey I Chizhik, Martin J Schnermann, Markus Sauer, Oleksii Nevskyi, Jörg Enderlein
ABSTRACT
Over the past two decades, super-resolution microscopy has seen a tremendous development in speed and resolution, but for most of its methods, there exists a remarkable gap between lateral and axial resolution, which is by a factor of 2 to 3 worse. One recently developed method to close this gap is metal-induced energy transfer (MIET) imaging, which achieves an axial resolution down to nanometers. It exploits the distance-dependent quenching of fluorescence when a fluorescent molecule is brought close to a metal surface. In the present manuscript, we combine the extreme axial resolution of MIET imaging with the extraordinary lateral resolution of single-molecule localization microscopy, in particular with direct stochastic optical reconstruction microscopy (dSTORM). This combination allows us to achieve isotropic three-dimensional super-resolution imaging of subcellular structures. Moreover, we used spectral demixing for implementing dual-color MIET-dSTORM that allows us to image and colocalize, in three dimensions, two different cellular structures simultaneously.
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder