- Use of recombinant antigens as diagnostic reagents in enzyme-linked immunosorbent assays.
Use of recombinant antigens as diagnostic reagents in enzyme-linked immunosorbent assays.
Methods in molecular medicine (1998-01-01)
A Warnes, A R Fooks, J R Stephenson
PMID21390846
ABSTRACT
The introduction of genetic engineering techniques has allowed the controlled and efficient production of recombinant proteins. This presents scientists with the opportunity to use a wide range of proteins for a number purposes, previously unavailable because of problems relating to expression, purification, or stabihty when considering the use of native proteins.
Sigma-Aldrich
Anti-Human IgG4−Biotin antibody, Mouse monoclonal, clone HP-6025, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG2−Biotin antibody, Mouse monoclonal, clone HP-6014, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG1−Biotin antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG3−Biotin antibody, Mouse monoclonal, clone HP-6050, purified from hybridoma cell culture