Merck
Preparation and Characterization of Tau Oligomer Strains.
Methods in molecular biology (Clifton, N.J.) (2018-06-11)
Urmi Sengupta, Mariana Carretero-Murillo, Rakez Kayed
ABSTRACT
An increasing number of studies have demonstrated the existence of multiple conformational entities of tau, as have been observed for prion protein. We have developed and optimized techniques to isolate and study oligomeric tau strains both in vitro and ex vivo. Moreover, we have modified protocols that demonstrate the seeding properties of oligomeric tau strains that are capable of propagating in vivo. These methods and protocols are explained in this chapter.
Sigma-Aldrich
Anti-cleaved-Tau (Asp421) Antibody, clone C3, clone C3, Upstate®, from mouse