Merck
  • Dimethylarginine dimethylaminohydrolase 1 as a novel regulator of oligodendrocyte differentiation in the central nervous system remyelination.
Dimethylarginine dimethylaminohydrolase 1 as a novel regulator of oligodendrocyte differentiation in the central nervous system remyelination.
Glia (2021-07-17)
Akiko Uyeda, Lili Quan, Yuki Kato, Nagaaki Muramatsu, Shogo Tanabe, Kazuhisa Sakai, Noritaka Ichinohe, Yukio Kawahara, Tatsunori Suzuki, Rieko Muramatsu
ABSTRACT
Remyelination is a regenerative process that restores the lost neurological function and partially depends on oligodendrocyte differentiation. Differentiation of oligodendrocytes spontaneously occurs after demyelination, depending on the cell intrinsic mechanisms. By combining a loss-of-function genomic screen with a web-resource-based candidate gene identification approach, we identified that dimethylarginine dimethylaminohydrolase 1 (DDAH1) is a novel regulator of oligodendrocyte differentiation. Silencing DDAH1 in oligodendrocytes prevented the expression of myelin basic protein in mouse oligodendrocyte culture with the change in expression of genes annotated with oligodendrocyte development. DDAH1 inhibition attenuated spontaneous remyelination in a cuprizone-induced demyelinated mouse model. Conversely, increased DDAH1 expression enhanced remyelination capacity in experimental autoimmune encephalomyelitis. These results provide a novel therapeutic option for demyelinating diseases by modulating DDAH1 activity.
Sigma-Aldrich
Anti-Aspa/Nur7 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse