Merck
  • African Swine Fever Virus Regulates Host Energy and Amino Acid Metabolism To Promote Viral Replication.
African Swine Fever Virus Regulates Host Energy and Amino Acid Metabolism To Promote Viral Replication.
Journal of virology (2021-12-16)
Qiao Xue, Huisheng Liu, Zixiang Zhu, Fan Yang, Yingying Song, Zongqiang Li, Zhaoning Xue, Weijun Cao, Xiangtao Liu, Haixue Zheng
ABSTRACT
African swine fever is one of the most serious viral diseases caused by African swine fever virus (ASFV). The metabolic changes induced by ASFV infection remain unknown. Here, porcine alveolar macrophages (PAMs) infected with ASFV was analyzed by ultrahigh-performance liquid chromatography-quadrupole time-of-flight tandem mass spectrometry (UHPLC-QTOF-MS) in combination with multivariate statistical analysis. A total of 90 metabolites were significantly changed after ASFV infection, and most of them were amino acids and tricarboxylic acid (TCA) cycle intermediates. ASFV infection induced an increase in most of amino acids in the host during the early stages of infection, and amino acids decreased in the late stages of infection. ASFV infection did not significantly affect the glycolysis pathway, whereas it induced increases in citrate, succinate, α-ketoglutarate, and oxaloacetate levels in the TCA cycle, suggesting that ASFV infection promoted the TCA cycle. The activities of aspartate aminotransferase and glutamate production were significantly elevated in ASFV-infected cells and pigs, resulting in reversible transition between TCA cycle and amino acid synthesis. Aspartate, glutamate, and TCA cycle were essential for ASFV replication. In addition, ASFV infection induced an increase in lactate level using lactate dehydrogenase, which led to low expression of beta interferon (IFN-β) and increased ASFV replication. Our data, for the first time, indicate that ASFV infection controls IFN-β production through RIG-I-mediated signaling pathways. These data identified a novel mechanism evolved by ASFV to inhibit host innate immune responses and provide insights for development of new preventive or therapeutic strategies targeting the altered metabolic pathways. IMPORTANCE In order to promote viral replication, viruses often cause severe immunosuppression and seize organelles to synthesize a large number of metabolites required for self-replication. African swine fever virus (ASFV) has developed many strategies to evade host innate immune responses. However, the impact of ASFV infection on host cellular metabolism remains unknown. Here, for the first time, we analyzed the metabolomic profiles of ASFV-infected PAMs. ASFV infection increased host TCA cycle and amino acid metabolism. Aspartate, glutamate, and TCA cycle promoted ASFV replication. ASFV infection also induced the increase of lactate production to inhibit innate immune responses for self-replication. This study identified novel immune evasion mechanisms utilized by ASFV and provided insights into ASFV-host interactions, which is critical for guiding the design of new prevention strategies against ASFV targeting the altered metabolic pathways.