Merck
  • AS602801, an Anti-Cancer Stem Cell Drug Candidate, Suppresses Gap-junction Communication Between Lung Cancer Stem Cells and Astrocytes.
AS602801, an Anti-Cancer Stem Cell Drug Candidate, Suppresses Gap-junction Communication Between Lung Cancer Stem Cells and Astrocytes.
Anticancer research (2018-09-09)
Kenta Kuramoto, Masahiro Yamamoto, Shuhei Suzuki, Tomomi Sanomachi, Keita Togashi, Shizuka Seino, Chifumi Kitanaka, Masashi Okada
ABSTRACT
Cancer stem cells (CSCs) are associated with tumorigenesis, recurrence, and metastasis. Cell-cell communication via gap junctions (GJs) between metastatic cancer cells and astrocytes is necessary for brain metastasis. Agents targeting communication between CSCs and astrocytes are expected to suppress brain metastasis. Using the A549 CSC, a cancer stem-like cell derived from A549, we examined the effect of AS602801, an anti-cancer stem cell agent whose safety has been confirmed in a phase 2 clinical trial, on GJ communication and connexin expression using a dye-transfer assay and immunoblot analysis, respectively. AS602801 specifically suppressed cell-cell communication in A549 CSCs without any suppression of GJ communication in astrocytes; it also decreased the expression of connexin 43, a constituent of GJs, in A549 CSCs. The anti-cancer stem cell agent, AS602801, is a potential drug candidate against brain metastasis.
Sigma-Aldrich
AS602801, ≥98% (HPLC)