Merck
  • A novel form of integrin dysfunction involving beta1, beta2, and beta3 integrins.
A novel form of integrin dysfunction involving beta1, beta2, and beta3 integrins.
The Journal of clinical investigation (2003-01-04)
Alison McDowall, David Inwald, Birgit Leitinger, Alison Jones, Ri Liesner, Nigel Klein, Nancy Hogg
ABSTRACT
The adhesion receptors known as integrins perform key functions for hematopoietic cells. The platelet integrin alphaIIbbeta3 is critical in hemostasis, and the beta1 and beta2 integrins on leukocytes have many roles in cell-mediated immunity. Mutations in the beta2 subunit lead to integrin nonexpression and to an immune deficiency, leukocyte adhesion deficiency-1. Mutations in either the alpha or beta subunit of alphaIIbbeta3 usually lead to integrin nonexpression and a bleeding tendency termed Glanzmann thrombasthenia. Here we describe a unique patient with clinical features of both Glanzmann thrombasthenia and leukocyte adhesion deficiency-1. The patient has normal expression of beta1, beta2, and beta3 integrins, but all are dysfunctional. The key findings are that "inside-out" signaling pathways leading to integrin activation are defective and that this is associated with abnormal integrin clustering. The integrins themselves are intact and capable of function following extracellular stimulation. T cell motility is normal, as are the expression levels and electrophoretic characteristics of all cytoskeletal and signaling proteins tested, except PKC-alpha, which has enhanced expression in the patient's cells. To our knowledge, this is the first description of a dysfunction affecting three classes of integrins. We propose that it is caused by a lesion in an intracellular factor or signaling pathway essential for integrin activation in hematopoietic cells and results in lack of regulation of clustering, an essential component of integrin-mediated adhesion.
Sigma-Aldrich
Anti-Goat/Sheep IgG−FITC antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone B3B11, clone B3B11, Chemicon®, from mouse